Ethyl ethoxyacetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 2-ethoxyacetate, Ethoxyacetic acid ethyl ester

M-Clarity™ quality level = MQ200

374172-10G 374172-50G
SIAL374172-10GEA 122 GBP
SIAL374172-10G SIAL374172-50G
Ethyl ethoxyacetate, Sigma-Aldrich®
Ethyl ethoxyacetate
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 156…157 °C (1013 hPa)
Density: 0.9996 g/cm³ (0 °C)
MDL Number: MFCD00040492
CAS Number: 817-95-8
EINECS: 212-447-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR