Benzyl acetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B15805-100G B15805-1KG
SIALB15805-100GEA 47.5 GBP
SIALB15805-100G SIALB15805-1KG
Benzyl acetate, Sigma-Aldrich®
Benzyl acetate
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: –51 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00008712
CAS Number: 140-11-4
EINECS: 205-399-7
Merck Index: 13,01125

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR