2-Bromoethyl acetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Acetic acid (2-bromoethyl) ester

137685-100G 137685-500G 137685-5G
SIAL137685-100GEA 99.6 GBP
SIAL137685-100G SIAL137685-500G SIAL137685-5G
2-Bromoethyl acetate, Sigma-Aldrich®
2-Bromoethyl acetate
Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –14 °C
Density: 1,4623 g/cm³ (20 °C)
MDL Number: MFCD00000235
CAS Number: 927-68-4
EINECS: 213-159-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR