Dimethyldiphenyltin

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Dimethyldiphenylstannane

39617.14EA 484 GBP
39617.14 39617.03 39617.06
Dimethyldiphenyltin
Dimethyldiphenyltin
Formula: C₁₄H₁₆Sn
Storage Temperature: Ambient
MDL Number: MFCD00058812
CAS Number: 1080-43-9
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR