Decacarbonyldirhenium ≥98%

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Rhenium carbonyl

STRE75-1800-5GEA 0 GBP
STRE75-1800-5G
Decacarbonyldirhenium ≥98%
Decacarbonyldirhenium

Metal Carbonyls, Volatile Metal Carbonyls for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀O₁₀Re₂
MW: 652.52 g/mol
Melting Pt: 170 °C (dec.)
Density: 2.87
Storage Temperature: Ambient
MDL Number: MFCD00011198
CAS Number: 14285-68-8
EINECS: 238-202-8
UN: 3466
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR