Cytochalasin D

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-triene-1,17-dione 21(R)-acetate

SERA18017.01EA 0 GBP
SERA18017.02 SERA18017.01
Cytochalasin D
Cytochalasin D
Formula: C₃₀H₃₇NO₆
MW: 507.63 g/mol
Melting Pt: 259…260 °C
CAS Number: 22144-77-0
EINECS: 244-804-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR