Diaminomaleonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002905-25G 002905-100G
FLUO002905-25GEA 0 GBP
FLUO002905-25G FLUO002905-100G
Diaminomaleonitrile
Diaminomaleonitrile
Formula: C₄H₄N₄
MW: 108.1 g/mol
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 178…181 °C
Density: 1.368 g/cm³ (20 °C)
MDL Number: MFCD00001870
CAS Number: 1187-42-4
EINECS: 214-697-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR