2-Indolinone
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone
IN-0069-5G
IN-0069-25G
IN-0069-100G
COBBIN-0069-5GEA
0
GBP
COBBIN-0069-5G
COBBIN-0069-25G
COBBIN-0069-100G
2-Indolinone
2-Indolinone
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 313 °C (1013 hPa) Melting Pt: 122…125 °C Density: 1.32 g/cm³ (20 °C) |
MDL Number:
MFCD00005711 CAS Number: 59-48-3 EINECS: 200-429-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...