Dexamethasone
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Dexamethasonum
, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
EHERC12170400EA
130
GBP
EHERC12170400
Dexamethasone
Dexamethasone
Formula:
C₂₂H₂₉FO₅ MW: 392.47 g/mol Melting Pt: 268…271 °C |
MDL Number:
MFCD00064136 CAS Number: 50-02-2 EINECS: 200-003-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...