6-Methylindole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
6-Methyl-1H-indole
IN-0032-1G
IN-0032-5G
IN-0032-25G
COBBIN-0032-1GEA
0
GBP
COBBIN-0032-1G
COBBIN-0032-5G
COBBIN-0032-25G
6-Methylindole
6-Methylindole
Formula:
C₉H₉N MW: 131.18 g/mol Boiling Pt: 260 °C (1013 hPa) Melting Pt: 30…31 °C Density: 1.05 g/cm³ (20 °C) |
MDL Number:
MFCD00005682 CAS Number: 3420-02-8 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...