6-Fluoroindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Fluoro-1H-indole

IN-0052-5G IN-0052-25G
COBBIN-0052-5GEA 0 GBP
COBBIN-0052-5G COBBIN-0052-25G
6-Fluoroindole
6-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00056933
CAS Number: 399-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR