Indole-6-carbonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Cyano-1H-indole, 6-Cyanoindole, 1H-Indole-6-carbonitrile

IN-0007-5G IN-0007-1G IN-0007-25G
COBBIN-0007-5GEA 0 GBP
COBBIN-0007-5G COBBIN-0007-1G COBBIN-0007-25G
Indole-6-carbonitrile
Indole-6-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
MDL Number: MFCD00016732
CAS Number: 15861-36-6
EINECS: 239-988-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR