4-Fluoro-2-methylbenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Fluoro-2-methylbenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ100
558567-1G
558567-5G
SIAL558567-1GEA
35.8
GBP
SIAL558567-1G
SIAL558567-5G
4-Fluoro-2-methylbenzenesulphonyl chloride, Sigma-Aldrich®
4-Fluoro-2-methylbenzenesulphonyl chloride
Formula:
C₇H₆ClFO₂S MW: 208.64 g/mol Boiling Pt: 245…246 °C (1013 hPa) Density: 1.433 g/cm³ (20 °C) |
MDL Number:
MFCD03094390 CAS Number: 7079-48-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...