4-Bromothiobenzamide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

684732-1G 684732-5G
SIAL684732-1GEA 48.2 GBP
SIAL684732-1G SIAL684732-5G
4-Bromothiobenzamide, Sigma-Aldrich®
4-Bromothiobenzamide
Formula: C₇H₆BrNS
MW: 216.1 g/mol
MDL Number: MFCD04114360
CAS Number: 26197-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR