4-Bromothiazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1436-5G
HI-1436-1G
COBBHI-1436-5GEA
0
GBP
COBBHI-1436-5G
COBBHI-1436-1G
4-Bromothiazole
4-Bromothiazole
Formula:
C₃H₂BrNS MW: 164.03 g/mol |
MDL Number:
MFCD06657592 CAS Number: 34259-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...