4-Bromo-2-methoxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7176-1G PY-7176-5G
COBBPY-7176-1GEA 0 GBP
COBBPY-7176-1G COBBPY-7176-5G
4-Bromo-2-methoxypyridine
4-Bromo-2-methoxypyridine
Formula: C₆H₆BrNO
MDL Number: MFCD08277268
CAS Number: 100367-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR