4-Bromo-2-fluorobenzoic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4096-5G
CA-4096-100G
CA-4096-25G
COBBCA-4096-5GEA
0
GBP
COBBCA-4096-5G
COBBCA-4096-100G
COBBCA-4096-25G
4-Bromo-2-fluorobenzoic acid
4-Bromo-2-fluorobenzoic acid
Formula:
C₇H₄BrFO₂ MW: 219.01 g/mol |
MDL Number:
MFCD00143263 CAS Number: 112704-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...