Acacetin, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5,7-Dihydroxy-2-(4-methoxyphenyl)chromen-4-one, Linarisenin, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one, 4'-Methoxy-5,7-dihydroxyflavone, 5,7-Dihydroxy-4'-methoxyflavone, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-methoxyphenyl)-4-benzopyrone, Linarigenin, Buddleoflavonol, 4'-Methoxyapigenin

M-Clarity™ quality level = MQ100

00017-100MG 00017-25MG
SIAL00017-100MGEA 1190 GBP
SIAL00017-100MG SIAL00017-25MG
Acacetin, Sigma-Aldrich®
Acacetin
Formula: C₁₆H₁₂O₅
MW: 284.27 g/mol
MDL Number: MFCD00016936
CAS Number: 480-44-4
EINECS: 207-552-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR