4-Acetoxybenzaldehyde

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Ethanoyloxy)benzaldehyde, 4-Formylphenyl acetate

A1170.10 A1170.50
XXXXA1170.10EA 0 GBP
XXXXA1170.10 XXXXA1170.50
4-Acetoxybenzaldehyde
4-Acetoxybenzaldehyde
Formula: C₉H₈O₃
MW: 164.16 g/mol
MDL Number: MFCD00003384
CAS Number: 878-00-2
EINECS: 212-898-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR