3-Iodo-4-methoxybenzoic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Iodo-p-anisic acid
HC-3319-5G
HC-3319-25G
HC-3319-1G
COBBHC-3319-5GEA
0
GBP
COBBHC-3319-5G
COBBHC-3319-25G
COBBHC-3319-1G
3-Iodo-4-methoxybenzoic acid
3-Iodo-4-methoxybenzoic acid
Formula:
C₈H₇IO₃ MW: 278.05 g/mol |
MDL Number:
MFCD00017347 CAS Number: 68507-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...