5-Fluoro-2-hydroxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7328-1G PY-7328-25G PY-7328-5G
COBBPY-7328-1GEA 0 GBP
COBBPY-7328-1G COBBPY-7328-25G COBBPY-7328-5G
5-Fluoro-2-hydroxypyridine
5-Fluoro-2-hydroxypyridine
Formula: C₅H₄FNO
MW: 113.09 g/mol
MDL Number: MFCD03092918
CAS Number: 51173-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR