3-Fluoro-2-hydroxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7344-1G
PY-7344-25G
PY-7344-5G
COBBPY-7344-1GEA
0
GBP
COBBPY-7344-1G
COBBPY-7344-25G
COBBPY-7344-5G
3-Fluoro-2-hydroxypyridine
3-Fluoro-2-hydroxypyridine
Formula:
C₅H₄FNO MW: 113.09 g/mol |
MDL Number:
MFCD03095042 CAS Number: 1547-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...