2-Fluoronicotinonitrile ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Cyano-2-fluoropyridine
L19538.03
L19538.06
L19538.03EA
27.7
GBP
L19538.03
L19538.06
2-Fluoronicotinonitrile ≥98%
2-Fluoronicotinonitrile
Purity: 98%
CAS No.: 3939-13-7
Molecular Formula: C6H3FN2
Formula Weight: 122.10
Melting Point: 27-30°
Beilstein Reference: 386567
Formula:
C₆H₃FN₂ MW: 122.1 g/mol Boiling Pt: 196 °C (1013 hPa) Melting Pt: 27…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095082 CAS Number: 3939-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...