3-Chloropyridine
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBOS-7451-25GEA
0
GBP
COBBOS-7451-25G
COBBOS-7451-5G
COBBOS-7451-100G
3-Chloropyridine
3-Chloropyridine
Formula:
C₅H₄ClN MW: 113.55 g/mol Boiling Pt: 148 °C (1013 hPa) Melting Pt: –0.5 °C Density: 1.194 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006375 CAS Number: 626-60-8 EINECS: 210-955-7 UN: 1100 ADR: 3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...