3-Chloro-4-methylbenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chlor-4-methylbenzenesulphonyl chloride, 2-Chloro-p-toluenesulphochloride, 3-Chloro-4-methylbenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

554243-1G 554243-5G
SIAL554243-1GEA 94.5 GBP
SIAL554243-1G SIAL554243-5G
3-Chloro-4-methylbenzenesulphonyl chloride, Sigma-Aldrich®
3-Chloro-4-methylbenzenesulphonyl chloride
Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
MDL Number: MFCD00203987
CAS Number: 42413-03-6
EINECS: 412-890-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR