3-Bromobenzenesulphonyl chloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7484-5G OS-7484-25G OS-7484-100G
COBBOS-7484-5GEA 0 GBP
COBBOS-7484-5G COBBOS-7484-25G COBBOS-7484-100G
3-Bromobenzenesulphonyl chloride
3-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Boiling Pt: 95…95.5 °C (0.8 mmHg)
Melting Pt: 31…33 °C
Density: 1.773 g/cm³ (20 °C)
MDL Number: MFCD00052313
CAS Number: 2905-24-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR