5-Bromo-2-fluoropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7039-100G
PY-7039-25G
PY-7039-5G
COBBPY-7039-100GEA
0
GBP
COBBPY-7039-100G
COBBPY-7039-25G
COBBPY-7039-5G
5-Bromo-2-fluoropyridine
5-Bromo-2-fluoropyridine
Formula:
C₅H₃BrFN MW: 175.99 g/mol Boiling Pt: 63 °C (15 mmHg) |
MDL Number:
MFCD01863742 CAS Number: 766-11-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...