3-Bromo-5-fluoropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7127-1G PY-7127-25G PY-7127-5G
COBBPY-7127-1GEA 0 GBP
COBBPY-7127-1G COBBPY-7127-25G COBBPY-7127-5G
3-Bromo-5-fluoropyridine
3-Bromo-5-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
MDL Number: MFCD04112555
CAS Number: 407-20-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR