3-Amino-4-iodopyridine 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Iodo-3-pyridylamine, 4-Iodopyridin-3-amine, 4-Iodo-pyridin-3-ylamine

H50023.03 H50023.06
H50023.03EA 113 GBP
H50023.03 H50023.06
3-Amino-4-iodopyridine 98%
3-Amino-4-iodopyridine

Purity: 98% CAS No.: 105752-11-2 Molecular Formula: C5H5IN2 Formula Weight: 220.01 Melting Point: 55-68° MDL No.: MFCD04971334

Formula: C₅H₅IN₂
MW: 220.01 g/mol
Melting Pt: 55…68 °C
Storage Temperature: Ambient
MDL Number: MFCD04971334
CAS Number: 105752-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR