3,6-Dimethoxypyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2808-5G HC-2808-1G
COBBHC-2808-5GEA 0 GBP
COBBHC-2808-5G COBBHC-2808-1G
3,6-Dimethoxypyridazine
3,6-Dimethoxypyridazine
Formula: C₆H₈N₂O₂
MDL Number: MFCD00234224
CAS Number: 4603-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR