3,5-Bis(trifluoromethyl)benzamide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
559997-1G
559997-5G
SIAL559997-1GEA
31.5
GBP
SIAL559997-1G
SIAL559997-5G
3,5-Bis(trifluoromethyl)benzamide, Sigma-Aldrich®
3,5-Bis(trifluoromethyl)benzamide
Formula:
C₉H₅F₆NO MW: 257.14 g/mol |
MDL Number:
MFCD00015511 CAS Number: 22227-26-5 EINECS: 244-849-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...