4,5-Difluoro-2-nitrobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4641-5G CA-4641-25G CA-4641-1G
COBBCA-4641-5GEA 0 GBP
COBBCA-4641-5G COBBCA-4641-25G COBBCA-4641-1G
4,5-Difluoro-2-nitrobenzoic acid
4,5-Difluoro-2-nitrobenzoic acid
Formula: C₇H₃F₂NO₄
MW: 203.1 g/mol
MDL Number: MFCD00799521
CAS Number: 20372-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR