3,4-Dibromopyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7723-5G OS-7723-25G OS-7723-1G
COBBOS-7723-5GEA 0 GBP
COBBOS-7723-5G COBBOS-7723-25G COBBOS-7723-1G
3,4-Dibromopyridine
3,4-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
MDL Number: MFCD00234016
CAS Number: 13534-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR