Ethyl 3,4-diaminobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Diaminobenzoic acid ethyl ester, Ethyl 3,4-diaminobenzoate

020165-5G 020165-25G
FLUO020165-5GEA 0 GBP
FLUO020165-5G FLUO020165-25G
Ethyl 3,4-diaminobenzoate
Ethyl 3,4-diaminobenzoate
Formula: C₉H₁₂N₂O₂
MW: 180.21 g/mol
MDL Number: MFCD00179337
CAS Number: 37466-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR