3-Acetoxybenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Formylphenyl acetate

002484-5G 002484-25G
FLUO002484-5GEA 0 GBP
FLUO002484-5G FLUO002484-25G
3-Acetoxybenzaldehyde
3-Acetoxybenzaldehyde
Formula: C₉H₈O₃
MW: 164.16 g/mol
MDL Number: MFCD00016603
CAS Number: 34231-78-2
EINECS: 251-890-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR