2-Nitro-4-(trifluoromethyl)benzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Nitro-4-(trifluoromethyl)benzene-1-sulphonyl chloride

009304-5G 009304-25G
FLUO009304-5GEA 0 GBP
FLUO009304-5G FLUO009304-25G
2-Nitro-4-(trifluoromethyl)benzenesulphonyl chloride
2-Nitro-5-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃ClF₃NO₄S
MW: 289.62 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 42…44 °C
MDL Number: MFCD00013456
CAS Number: 837-95-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR