Methyl salicylate
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Methyl 2-hydroxybenzoate
, Wintergreen Oil
EHERC15143400EA
213
GBP
EHERC15143400
Methyl salicylate
Methyl salicylate
Formula:
2-(HO)C₆H₄CO₂CH₃ MW: 152.15 g/mol Boiling Pt: 432 °C (1013 hPa) Melting Pt: 100…101 °C Density: 1.184 g/cm³ (20 °C) Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002214 CAS Number: 119-36-8 EINECS: 204-317-7 Merck Index: 13,06143 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...