Furan-2-boronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Furan-2-ylboronic acid

010999-5G 010999-25G
FLUO010999-5GEA 0 GBP
FLUO010999-5G FLUO010999-25G
Furan-2-boronic acid
Furan-2-boronic acid
Formula: C₄H₅BO₃
MW: 111.89 g/mol
MDL Number: MFCD00799544
CAS Number: 13331-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR