2-Fluoro-3-pyridineboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Fluoro-3-pyridinylboronic acid, (2-Fluoropyridin-3-yl)boronic acid, 2-Fluoropyridine-3-boronic acid
BB-3476-5G
BB-3476-25G
COBBBB-3476-5GEA
0
GBP
COBBBB-3476-5G
COBBBB-3476-25G
2-Fluoro-3-pyridineboronic acid
2-Fluoro-3-pyridineboronic acid
Formula:
C₅H₅BFNO₂ MW: 140.91 g/mol |
MDL Number:
MFCD03411557 CAS Number: 174669-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...