2-Fluoro-3-iodopyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

SS-3009-1G SS-3009-25G SS-3009-5G
COBBSS-3009-1GEA 0 GBP
COBBSS-3009-1G COBBSS-3009-25G COBBSS-3009-5G
2-Fluoro-3-iodopyridine
2-Fluoro-3-iodopyridine
Formula: C₅H₃FIN
MW: 222.99 g/mol
MDL Number: MFCD03095287
CAS Number: 113975-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR