2-Chloroquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PY-7806-5G
PY-7806-100G
PY-7806-25G
COBBPY-7806-5GEA
0
GBP
COBBPY-7806-5G
COBBPY-7806-100G
COBBPY-7806-25G
2-Chloroquinoline
2-Chloroquinoline
Formula:
C₉H₆ClN MW: 163.61 g/mol Boiling Pt: 266…267 °C (1013 hPa) Melting Pt: 38 °C Density: 1,2464 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006741 CAS Number: 612-62-4 EINECS: 210-317-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...