2-Chloro-3-(trifluoromethyl)pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7569-25G
OS-7569-5G
OS-7569-100G
COBBOS-7569-25GEA
0
GBP
COBBOS-7569-25G
COBBOS-7569-5G
COBBOS-7569-100G
2-Chloro-3-(trifluoromethyl)pyridine
2-Chloro-3-(trifluoromethyl)pyridine
Formula:
C₆H₃ClF₃N MW: 181.54 g/mol Boiling Pt: 75…76 °C (30 mmHg) Melting Pt: 38…39 °C |
MDL Number:
MFCD00042223 CAS Number: 65753-47-1 EINECS: 424-520-6 UN: 2923 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...