2-Bromothiobenzamide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

684791-1G 684791-5G
SIAL684791-1GEA 75.9 GBP
SIAL684791-1G SIAL684791-5G
2-Bromothiobenzamide, Sigma-Aldrich®
2-Bromothiobenzamide
Formula: C₇H₆BrNS
MW: 216.1 g/mol
MDL Number: MFCD04973314
CAS Number: 30216-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR