2-Bromo-6-fluorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7682-5G OS-7682-25G OS-7682-1G
COBBOS-7682-5GEA 0 GBP
COBBOS-7682-5G COBBOS-7682-25G COBBOS-7682-1G
2-Bromo-6-fluorobenzoic acid
2-Bromo-6-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD01569539
CAS Number: 2252-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR