2-Bromo-5-chloropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7032-25G PY-7032-5G PY-7032-100G
COBBPY-7032-25GEA 11.87 GBP
COBBPY-7032-25G COBBPY-7032-5G COBBPY-7032-100G
2-Bromo-5-chloropyridine
2-Bromo-5-chloropyridine
Formula: C₅H₃BrClN
MDL Number: MFCD00234006
CAS Number: 40473-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR