2-Bromo-4-methylimidazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-methyl-1H-imidazole

HI-1178-5G HI-1178-1G
COBBHI-1178-5GEA 0 GBP
COBBHI-1178-5G COBBHI-1178-1G
2-Bromo-4-methylimidazole
2-Bromo-4-methylimidazole
Formula: C₄H₅BrN₂
MW: 161 g/mol
MDL Number: MFCD06659904
CAS Number: 23328-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR