2-Bromo-3-fluoropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7616-5G
OS-7616-25G
OS-7616-1G
COBBOS-7616-5GEA
0
GBP
COBBOS-7616-5G
COBBOS-7616-25G
COBBOS-7616-1G
2-Bromo-3-fluoropyridine
2-Bromo-3-fluoropyridine
Formula:
C₅H₃BrFN |
MDL Number:
MFCD04114128 CAS Number: 40273-45-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...