2-Bromo-3-(trifluoromethyl)pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7413-25G
OS-7413-5G
OS-7413-1G
OS-7413-100G
COBBOS-7413-25GEA
0
GBP
COBBOS-7413-25G
COBBOS-7413-5G
COBBOS-7413-1G
COBBOS-7413-100G
2-Bromo-3-(trifluoromethyl)pyridine
2-Bromo-3-(trifluoromethyl)pyridine
Formula:
C₆H₃BrF₃N MW: 226 g/mol |
MDL Number:
MFCD00153084 CAS Number: 175205-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...