2-Amino-5-iodopyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Iodopyridin-2-amine, 5-Iodo-2-pyridylamine

CA-4308-100G CA-4308-25G CA-4308-5G
COBBCA-4308-100GEA 0 GBP
COBBCA-4308-100G COBBCA-4308-25G COBBCA-4308-5G
2-Amino-5-iodopyridine
2-Amino-5-iodopyridine
Formula: C₅H₅IN₂
MW: 220.01 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 128…131 °C
MDL Number: MFCD00160312
CAS Number: 20511-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR