2-Amino-5-chloro-3-fluoropyridine 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Chloro-3-fluoro-2-pyridylamine, 5-Chloro-3-fluoropyridin-2-amine

H30135.03 H30135.06
H30135.03EA 79 GBP
H30135.03 H30135.06
2-Amino-5-chloro-3-fluoropyridine 98%
2-Amino-5-chloro-3-fluoropyridine

Purity: 98% CAS No.: 246847-98-3 Molecular Formula: C5H4ClFN2 Formula Weight: 146.55 Melting Point: 95-97° MDL No.: MFCD09878434

Formula: C₅H₄ClFN₂
MW: 146.55 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD09878434
CAS Number: 246847-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR